4-Nitrophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Warning

L11042-09 L11042-18 L11042-30
AAAL11042-09EA 55.4 USD
AAAL11042-09 AAAL11042-18 AAAL11042-30
4-Nitrophenyl isothiocyanate 97%
4-Nitrophenyl isothiocyanate

Purity: 97% CAS No.: 2131-61-5 Molecular Formula: C7H4N2O2S Formula Weight: 180.19 Odor: Irritating Melting Point: 105-110° Boiling Point: 137-138°/11mm Beilstein Reference: 640027 UN No.: 2923 MDL No.: MFCD00007307

Formula: C₇H₄N₂O₂S
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 108 - 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007307
CAS Number: 2131-61-5
EINECS: 218-359-9
UN: 2811
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR