2-Chloro-4-nitrophenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Chloro-1-isothiocyanato-4-nitrobenzene, 1-Chloro-2-isothiocyanato-5-nitrobenzene
L10898-03
L10898-06
AAAL10898-03EA
20.3
USD
AAAL10898-03
AAAL10898-06
2-Chloro-4-nitrophenyl isothiocyanate 97%
2-Chloro-4-nitrophenyl isothiocyanate
Purity: 97%
CAS No.: 23165-64-2
Molecular Formula: C7H3ClN2O2S
Formula Weight: 214.63
Odor: Irritating
Melting Point: 95-99°
Beilstein Reference: 2972987
UN No.: 2923
MDL No.: MFCD00041244
Formula:
C₇H₃ClN₂O₂S MW: 214.63 g/mol Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041244 CAS Number: 23165-64-2 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...