4-Bromo-2-chlorobenzonitrile 96%

Supplier: Thermo Scientific Chemicals
Danger

L10602-03 L10602-06
AAAL10602-03EA 17.7 USD
AAAL10602-03 AAAL10602-06
4-Bromo-2-chlorobenzonitrile 96%
4-Bromo-2-chlorobenzonitrile

Purity: 96% CAS No.: 154607-01-9 Molecular Formula: C7H3BrClN Formula Weight: 216.47 Melting Point: 67-72° Boiling Point: 142-143°/10mm UN No.: 3439 MDL No.: MFCD00040883

Formula: C₇H₃BrClN
MW: 216.46 g/mol
Boiling Pt: 142 - 143 °C (10 -rr)
Melting Pt: 67 - 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00040883
CAS Number: 154607-01-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR