2-Chloro-6-methylthiophene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-6-methylthiophenol

L10499-03 L10499-06 L10499-14
AAAL10499-03EA 49.3 USD
AAAL10499-03 AAAL10499-06 AAAL10499-14
2-Chloro-6-methylthiophene 96%
2-Chloro-6-methylthiophene

Purity: 96% CAS No.: 18858-05-4 Molecular Formula: C7H7ClS Formula Weight: 158.65 Refractive Index: 1.5950 Boiling Point: 135-136°/35mm MDL No.: MFCD00041422

Formula: C₅H₅ClS
MW: 132.61 g/mol
Boiling Pt: 135…136 °C/35 mm
Storage Temperature: Ambient
MDL Number: MFCD00041422
CAS Number: 18858-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR