(S)-(+)-1-Phenyl-1,2-ethanediol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-(+)-Phenyl-1,2-ethanediol, (S)-1-Phenylethane-1,2-diol, (S)-(+)-Phenylethane-1,2-diol, (S)-(+)-α,β-Dihydroxyethylbenzene
L09836-MD
L09836-03
AAAL09836-MDEA
31.6
USD
AAAL09836-MD
AAAL09836-03
(S)-(+)-1-Phenyl-1,2-ethanediol 97%
(S)-(+)-1-Phenyl-1,2-ethanediol
Purity: 97%
CAS No.: 25779-13-9
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 64-66°
Boiling Point: 272-274°
MDL No.: MFCD00066256
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066256 CAS Number: 25779-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...