1,2-Dimethylnaphthalene 98%

Supplier: Thermo Scientific Chemicals

L09774-03 L09774-MD
AAAL09774-03EA 77.87 USD
AAAL09774-03 AAAL09774-MD
1,2-Dimethylnaphthalene 98%
1,2-Dimethylnaphthalene

Purity: 98% CAS No.: 573-98-8 Molecular Formula: C12H12 Formula Weight: 156.23 Flash Point: 110°(230°F) Refractive Index: 1.6160 Odor: Characteristic Boiling Point: 266-267° Beilstein Reference: 2039376 MDL No.: MFCD00004035

Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 266…267 °C
Density: 1.0219 g/cm³ (16.3 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004035
CAS Number: 573-98-8
EINECS: 209-364-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR