2,4-Dimethoxyphenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Isocyanato-2,4-dimethoxybenzene

L09762-03 L09762-06
AAAL09762-03EA 24.77 USD
AAAL09762-03 AAAL09762-06
2,4-Dimethoxyphenyl isocyanate 97%
2,4-Dimethoxyphenyl isocyanate

Purity: 97% CAS No.: 84370-87-6 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5475 Melting Point: 31-32° Boiling Point: 139-141°/11mm Beilstein Reference: 1106649 UN No.: 2811 MDL No.: MFCD00002005

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Boiling Pt: 139…141 °C/11 mm
Melting Pt: 31…32 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002005
CAS Number: 84370-87-6
EINECS: 282-747-4
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR