Dimethyl disulfide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimethyl disulphide, Methyl disulphide

L09679-0B L09679-30
AAAL09679-0BEA 86.6 USD
AAAL09679-0B AAAL09679-30
Dimethyl disulfide 99%
Dimethyl disulphide

Purity: 99% CAS No.: 624-92-0 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 10°(50°F) Refractive Index: 1.5253 Odor: Stench Melting Point: -85° Boiling Point: 108-109° Beilstein Reference: 1730824 UN No.: 2381 MDL No.: MFCD00008561

Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 108…109 °C
Melting Pt: –85 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008561
CAS Number: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR