4-Chlorobenzyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 97%
CAS No.: 3694-45-9
Molecular Formula: C8H6ClNS
Formula Weight: 183.66
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6145
Odor: Irritating
Boiling Point: 169-170°/25mm
Beilstein Reference: 2085917
UN No.: 2922
MDL No.: MFCD00041116
Formula:
C₈H₆ClNS Boiling Pt: 169…170 °C/25 mm Density: 1.270 Storage Temperature: Refrigerator |
MDL Number:
MFCD00041116 CAS Number: 3694-45-9 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...