(S)-(+)-2-Pentanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(+)-Propyl methyl carbinol, (S)-(+)-Methyl propyl carbinol, (S)-(+)-2-Hydroxy pentane
L09314-MD
L09314-03
AAAL09314-MDEA
81.8
USD
AAAL09314-MD
AAAL09314-03
(S)-(+)-2-Pentanol 97%
(S)-(+)-2-Pentanol
Purity: 97%
CAS No.: 26184-62-3
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 33°(91°F)
Refractive Index: 1.4060
Boiling Point: 118-119°
Beilstein Reference: 1718820
Optical Rotation: +14° (neat)
UN No.: 1105
MDL No.: MFCD00065952
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 118…119 °C Melting Pt: –50 °C Density: 0.810 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00065952 CAS Number: 26184-62-3 UN: 1105 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...