1,4-Butane diisothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Butylene diisothiocyanate

L09234-06 L09234-03
AAAL09234-06EA 187 USD
AAAL09234-06 AAAL09234-03
1,4-Butane diisothiocyanate 98%
1,4-Butane diisothiocyanate

Purity: 98% CAS No.: 4430-51-7 Molecular Formula: SCN(CH2)4NCS Formula Weight: 172.27 Odor: Irritating Melting Point: 30-33° Boiling Point: 170°/11mm Beilstein Reference: 1757138 UN No.: 2923 MDL No.: MFCD00041136

Formula: C₆H₈N₂S₂
MW: 172.27 g/mol
Boiling Pt: 170 °C (11 mmHg)
Melting Pt: 124
Storage Temperature: Ambient
MDL Number: MFCD00041136
CAS Number: 4430-51-7
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR