(±)-6-Methyl-5-hepten-2-ol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L09181-14
L09181-06
AAAL09181-14EA
107
USD
AAAL09181-14
AAAL09181-06
(±)-6-Methyl-5-hepten-2-ol 98%
(±)-6-Methyl-5-hepten-2-ol
Purity: 98%
CAS No.: 1569-60-4
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 67°(152°F)
Refractive Index: 1.4480
Boiling Point: 78-80°/15mm
Beilstein Reference: 1720073
MDL No.: MFCD00004561
Formula:
C₈H₁₆O Boiling Pt: 78…80 °C/15 mm Density: 0,84 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004561 CAS Number: 1569-60-4 EINECS: 216-377-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...