Methyl m-anisate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Methoxybenzoic acid methyl ester, 3-Anisic acid methyl ester, m-Anisic acid methyl ester, Methyl 3-methoxybenzoate

L09041-18 L09041-09
AAAL09041-18EA 145 USD
AAAL09041-18 AAAL09041-09
Methyl m-anisate 98%
Methyl m-anisate

Purity: 98% CAS No.: 5368-81-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Refractive Index: 1.5270 Boiling Point: 238° Beilstein Reference: 2208567 MDL No.: MFCD00017193

Formula: C₉H₁₀O₃
Boiling Pt: 238 °C
Density: 1,15
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017193
CAS Number: 5368-81-0
EINECS: 226-359-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR