4-Propoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

L08822-06 L08822-14
AAAL08822-06EA 31 USD
AAAL08822-06 AAAL08822-14
4-Propoxybenzaldehyde 97%
4-Propoxybenzaldehyde

Purity: 97% CAS No.: 5736-85-6 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 129-130°/9mm Beilstein Reference: 743408 MDL No.: MFCD00014134

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 129…130 °C/9 mm
Density: 1,039
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014134
CAS Number: 5736-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR