(S)-(+)-Hydratropic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Phenylpropanoic acid, (S)-(+)-2-Phenylpropionic acid, (S)-(+)-2-Phenylpropanoic acid

L08802-03 L08802-MD
AAAL08802-03EA 78.8 USD
AAAL08802-03 AAAL08802-MD
(S)-(+)-Hydratropic acid 97%
(S)-(+)-Hydratropic acid

Purity: 97% CAS No.: 7782-24-3 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: 30-34° Boiling Point: 264-266° Beilstein Reference: 2044507 Optical Rotation: +73° (c=1.6 in chloroform) MDL No.: MFCD00063139

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264…266 °C
Melting Pt: 30…34 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 148 °C(298 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063139
CAS Number: 7782-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR