L-Norvaline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

L08658-03 L08658-06
AAAL08658-03EA 34.8 USD
AAAL08658-03 AAAL08658-06
L-Norvaline 99%
L-Norvaline

Purity: 99% CAS No.: 6600-40-4 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 300° Beilstein Reference: 1721162 Merck Reference: 14,6716 Optical Rotation: +25° (c=10 in 20% HCl) MDL No.: MFCD00064421

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR