2-Amino-1,3-propanediol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol
L08426-14
L08426-06
AAAL08426-14EA
199
USD
AAAL08426-14
AAAL08426-06
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol
Purity: 98%
CAS No.: 534-03-2
Molecular Formula: C3H9NO2
Formula Weight: 91.11
Melting Point: 52-56°
Boiling Point: 128°/0.8mm
UN No.: 3259
MDL No.: MFCD00051487
Formula:
C₃H₉NO₂ MW: 91.11 g/mol Boiling Pt: 128 °C/0.8 mm Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051487 CAS Number: 534-03-2 EINECS: 208-584-0 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...