2,2,2-Trichloroethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,2-Trichloroethan-1-ol

L08163-14 L08163-22 L08163-06 L08163-36
AAL08163-14EA 61.24 USD
AAL08163-14 AAAL08163-22 AAL08163-06 AAL08163-36
2,2,2-Trichloroethanol 99%
2,2,2-Trichloroethanol

Purity: 99% CAS No.: 115-20-8 Molecular Formula: Cl3CCH2OH Formula Weight: 149.40 Flash Point: 110°(230°F) Refractive Index: 1.4895 Melting Point: 17-18° Boiling Point: 151-153° Beilstein Reference: 1697495 Merck Reference: 14,9638 MDL No.: MFCD00004677

Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
Boiling Pt: 151…153 °C
Melting Pt: 17…18 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004677
CAS Number: 115-20-8
EINECS: 204-071-0
ADR: 6.1,III
Merck Index: 12,09786

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR