Pyrene 98%

Supplier: Thermo Scientific Chemicals
Warning

L08162-22 L08162-14
AAAL08162-22EA 165.87 USD
AAAL08162-22 AAAL08162-14
Pyrene 98%
Pyrene

Purity: 98% CAS No.: 129-00-0 Molecular Formula: C16H10 Formula Weight: 202.26 Flash Point: 208°(406°F) Melting Point: 148-152° Boiling Point: 393° Beilstein Reference: 1307225 Merck Reference: 14,7963 MDL No.: MFCD00004136

Formula: C₁₆H₁₀
MW: 202.26 g/mol
Boiling Pt: 393 °C
Melting Pt: 148…152 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 208 °C (406 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004136
CAS Number: 129-00-0
EINECS: 204-927-3
UN: 3077
ADR: 9,III
Merck Index: 12,08147
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b03eb6b87b60315a282988993f066fd2d3af129&vid=d3f1ffa103d650d2e9f49139db04842dfa6ca0ff&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR