Ethyl-3-fluorobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-fluorobenzoate, 3-Fluorobenzoic acid ethyl ester

L07984-18 L07984-09
AAAL07984-18EA 69.1 USD
AAAL07984-18 AAAL07984-09
Ethyl-3-fluorobenzoate 98+%
Ethyl-3-fluorobenzoate

Purity: 98+% CAS No.: 451-02-5 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 82°(179°F) Refractive Index: 1.4850 Odor: Recognizable Boiling Point: 208° Beilstein Reference: 2045876 MDL No.: MFCD00000336

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 208 °C
Density: 1,136
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000336
CAS Number: 451-02-5
EINECS: 207-191-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR