1-Phenylpyrrole 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Phenylpyrrole, 1-Phenyl-1H-pyrrole
L07973-14
L07973-06
AAAL07973-14EA
141.07
USD
AAAL07973-14
AAAL07973-06
1-Phenylpyrrole 99%
1-Phenylpyrrole
Purity: 99%
CAS No.: 635-90-5
Molecular Formula: C10H9N
Formula Weight: 143.19
Melting Point: 57-60°
Boiling Point: 131°/30mm
MDL No.: MFCD00005343
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 131 °C/30 mm Melting Pt: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005343 CAS Number: 635-90-5 EINECS: 211-242-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...