2-Bromofluorene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07589-06
L07589-14
AAAL07589-06EA
50.84
USD
AAAL07589-06
AAAL07589-14
2-Bromofluorene 95%
2-Bromofluorene
Purity: 95%
CAS No.: 1133-80-8
Molecular Formula: C13H9Br
Formula Weight: 245.12
Melting Point: 110-114°
Boiling Point: 185°/135mm
MDL No.: MFCD00001115
Formula:
C₁₃H₉Br Boiling Pt: 350 °C (1013 hPa) Melting Pt: 102 - 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001115 CAS Number: 1133-80-8 EINECS: 214-480-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...