3-Fluorophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Fluorphenyl)hydrazin HCl, (3-Fluorophenyl)hydrazine hydrochloride, 3-Fluorophenylhydrazine hydrochloride, 1-(3-Fluorophenyl)hydrazine hydrochloride

L07418-06 L07418-14
AAAL07418-06EA 74.2 USD
AAAL07418-06 AAAL07418-14
3-Fluorophenylhydrazinium chloride 98%
3-Fluorophenylhydrazinium chloride

Purity: 98% CAS No.: 2924-16-5 Molecular Formula: C6H7FN2.HCl Formula Weight: 162.60 Melting Point: ca 268° dec. Beilstein Reference: 3627729

Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: ca 268 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012934
CAS Number: 2924-16-5
EINECS: 220-886-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR