1-Octanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 98%
CAS No.: 111-88-6
Molecular Formula: CH3(CH2)7SH
Formula Weight: 146.29
Flash Point: 68°(154°F)
Refractive Index: 1.4550
Melting Point: -49°
Boiling Point: 197-199°
Beilstein Reference: 1733101
UN No.: 3082
MDL No.: MFCD00004912
Formula:
C₈H₁₈S Boiling Pt: 197…199 °C Melting Pt: –49 °C Density: 0.843 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004912 CAS Number: 111-88-6 EINECS: 203-918-1 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...