3-(Trifluoromethyl)thiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol

L07191-06 L07191-03
AAAL07191-06EA 151 USD
AAAL07191-06 AAAL07191-03
3-(Trifluoromethyl)thiophenol 97%
3-(Trifluoromethyl)thiophenol

Purity: 97% CAS No.: 937-00-8 Molecular Formula: C7H5F3S Formula Weight: 178.18 Refractive Index: 1.4900 Odor: Stench Boiling Point: 71-72°/20mm Beilstein Reference: 1943114 UN No.: 2810 MDL No.: MFCD00041142

Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 161 - 163 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR