Dihexyl ether 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L07076-09
L07076-18
AAL07076-09EA
38.56
USD
AAL07076-09
AAL07076-18
Dihexyl ether 98%
Dihexyl ether
Purity: 98%
CAS No.: 112-58-3
Molecular Formula: C12H26O
Formula Weight: 186.34
Flash Point: 78°(172°F)
Refractive Index: 1.4204
Odor: Mild
Melting Point: -43°
Boiling Point: 228-230°
Beilstein Reference: 1738177
MDL No.: MFCD00009525
Formula:
C₁₂H₂₆O Boiling Pt: 223 °C (1013 hPa) Melting Pt: –43 °C Density: 0.8035 g/cm³ (20 °C) Flash Pt: 78 °C(172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009525 CAS Number: 112-58-3 EINECS: 203-987-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...