1-Phenyl-1-decanone 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Decanophenone, Decyl phenyl ketone, Nonyl phenyl ketone
L06923-06
L06923-14
AAAL06923-06EA
51.65
USD
AAAL06923-06
AAL06923-14
1-Phenyl-1-decanone 98+%
1-Phenyl-1-decanone
Purity: 98+%
CAS No.: 6048-82-4
Molecular Formula: C16H24O
Formula Weight: 232.37
Flash Point:
Formula:
C₆H₅C(O)(CH₂)₈CH₃ Boiling Pt: 165…167 °C/7 mm Melting Pt: 34…36 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009582 CAS Number: 6048-82-4 EINECS: 227-946-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...