Ethyl vinyl ketone 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Penten-3-one
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ≤ 0.1 %
L06829-06
L06829-14
AAAL06829-06EA
100.47
USD
AAAL06829-06
AAAL06829-14
Ethyl vinyl ketone 97% stabilized
Ethyl vinyl ketone
Purity: 97%
CAS No.: 1629-58-9
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: -6°(21°F)
Refractive Index: 1.4200
Odor: Pungent
Boiling Point: 103-104°
Beilstein Reference: 1735857
UN No.: 3286
MDL No.: MFCD00009316
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 103…104 °C Density: 0.8524 g/cm³ (15 °C) Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009316 CAS Number: 1629-58-9 EINECS: 216-624-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...