5-Amino-1H-indazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine

L06705-14 L06705-06
AAAL06705-14EA 173 USD
AAAL06705-14 AAL06705-06
5-Amino-1H-indazole 98%
5-Amino-1H-indazole

Purity: 98% CAS No.: 19335-11-6 Molecular Formula: C7H7N3 Formula Weight: 133.15 Odor: Odorless Melting Point: 172-178° Beilstein Reference: 3259 UN No.: 2811 MDL No.: MFCD00037975

Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 172…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00037975
CAS Number: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR