Cyclohexylméthanol 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L06538-14
L06538-22
AAAL06538-14EA
57.2
USD
AAAL06538-14
AAAL06538-22
Cyclohexylméthanol 99%
Cyclohexylméthanol
Purity: 99%
CAS No.: 100-49-2
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 71°(159°F)
Refractive Index: 1.4640
Boiling Point: 180-181°
Beilstein Reference: 773712
Merck Reference: 14,2731
MDL No.: MFCD00001510
Formula:
C₇H₁₄O Boiling Pt: 181 °C (1013 hPa) Melting Pt: -43 °C Density: 0.926 g/cm³ (20 °C) Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001510 CAS Number: 100-49-2 EINECS: 202-857-8 Merck Index: 13,02760 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...