4-Hydroxy-1-methylpiperidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Methyl-4-piperidinol, 1-Methyl-4-piperidinol, 1-Methylpiperidin-4-ol
L06415-36
L06415-22
AAAL06415-36EA
173
USD
AAAL06415-36
AAAL06415-22
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine
Purity: 98%
CAS No.: 106-52-5
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 87°(188°F)
Refractive Index: 1.4790
Melting Point: 26-30°
Boiling Point: 198-200°
Beilstein Reference: 103024
MDL No.: MFCD00006500
Formula:
C₆H₁₃NO Boiling Pt: 198…200 °C Melting Pt: 26…30 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006500 CAS Number: 106-52-5 EINECS: 203-406-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...