Octaphenylcyclotetrasiloxane 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2,4,4,6,6,8,8-Octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane

L06130-22 L06130-14
AAAL06130-22EA 154 USD
AAAL06130-22 AAAL06130-14
Octaphenylcyclotetrasiloxane 98+%
Octaphenylcyclotetrasiloxane

Purity: 98+% CAS No.: 546-56-5 Molecular Formula: C48H40O4Si4 Formula Weight: 793.19 Melting Point: 193-200° Boiling Point: 332°/1mm MDL No.: MFCD00003268

Formula: C₄₈H₄₀O₄Si₄
MW: 793.19 g/mol
Boiling Pt: 332 °C/1 mm
Melting Pt: 193…200 °C
Density: 1.089 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003268
CAS Number: 546-56-5
EINECS: 208-904-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR