Octyl-3,4,5-trihydroxybenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

L06081-14 L06081-22
AAL06081-14EA 60.48 USD
AAL06081-14 AAL06081-22
Octyl-3,4,5-trihydroxybenzoate 98+%
Octyl-3,4,5-trihydroxybenzoate

Purity: 98+% CAS No.: 1034-01-1 Molecular Formula: C15H22O5 Formula Weight: 282.34 Melting Point: 96-102° Beilstein Reference: 2132305

Formula: C₁₅H₂₂O₅
Melting Pt: 96…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002197
CAS Number: 1034-01-1
EINECS: 213-853-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR