Ethyl butyrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butanoic acid ethyl ester, Butyric acid ethyl ester
L06025-AU
L06025-AK
AAAL06025-AUEA
66.64
USD
AAAL06025-AU
AAAL06025-AK
Ethyl butyrate 99%
Ethyl butyrate
Purity: 99%
CAS No.: 105-54-4
Molecular Formula: CH3CH2CH2CO2CH2CH3
Formula Weight: 116.16
Flash Point: 26°(79°F)
Refractive Index: 1.3920
Melting Point: -93°
Boiling Point: 119-120°
Beilstein Reference: 506331
Merck Reference: 14,3775
UN No.: 1180
MDL No.: MFCD00009394
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 119…120 °C Melting Pt: –93 °C Density: 0.88 g/cm³ (20 °C) Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009394 CAS Number: 105-54-4 EINECS: 203-306-4 UN: 1180 ADR: 3,III Merck Index: 13,03810 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...