2-Chloro-6-methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

L05997-06 L05997-14
AAAL05997-06EA 23.9 USD
AAAL05997-06 AAAL05997-14
2-Chloro-6-methoxypyridine 98%
2-Chloro-6-methoxypyridine

Purity: 98% CAS No.: 17228-64-7 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Flash Point: 76°(168°F) Refractive Index: 1.5280 Boiling Point: 185-186° Beilstein Reference: 1364500 MDL No.: MFCD00006265

Formula: C₆H₆ClNO
Boiling Pt: 185…186 °C
Density: 1.207
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006265
CAS Number: 17228-64-7
EINECS: 241-264-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR