Methyl undecanoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05939-06
L05939-14
AAAL05939-06EA
35.6
USD
AAAL05939-06
AAAL05939-14
Methyl undecanoate 99%
Methyl undecanoate
Purity: 99%
CAS No.: 1731-86-8
Molecular Formula: CH3(CH2)9CO2CH3
Flash Point: 109°(228°F)
Refractive Index: 1.4290
Melting Point: -10°
Boiling Point: 246-248°
Beilstein Reference: 1762129
MDL No.: MFCD00008957
Formula:
C₁₂H₂₄O₂ Boiling Pt: 246…248 °C Melting Pt: –10 °C Density: 0.871 Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008957 CAS Number: 1731-86-8 EINECS: 217-053-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...