3,5-Diiodosalicylaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05783-09
L05783-18
AAAL05783-09EA
59.8
USD
AAAL05783-09
AAAL05783-18
3,5-Diiodosalicylaldehyde 98+%
3,5-Diiodosalicylaldehyde
Purity: 98+%
CAS No.: 2631-77-8
Molecular Formula: C7H4I2O2
Formula Weight: 373.92
Melting Point: 106-110°
Beilstein Reference: 1102552
Formula:
C₇H₄I₂O₂ Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003321 CAS Number: 2631-77-8 EINECS: 220-117-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...