2-Phenoxyaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-2-phenoxybenzol

L05558-22 L05558-14
AAAL05558-22EA 114 USD
AAAL05558-22 AAAL05558-14
2-Phenoxyaniline 98%
2-Phenoxyaniline

Purity: 99% CAS No.: 2688-84-8 Molecular Formula: C12H11NO Formula Weight: 185.23 Flash Point: 110°(230°F) Melting Point: 46-48° Boiling Point: 170°/18mm Beilstein Reference: 777766 MDL No.: MFCD00035765

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 170 °C/18 mm
Melting Pt: 46…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035765
CAS Number: 2688-84-8
EINECS: 220-254-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR