Molecular sieve A4 (0.4 nm, 4 Å), beads 1-2 mm (0.04-0.08 in)

Supplier: Thermo Scientific Chemicals

L05454-0B L05454-0I L05454-30
AAAL05454-0BEA 116 USD
AAAL05454-0B AAAL05454-0I AAAL05454-30
Molecular sieve A4 (0.4 nm, 4 Å), beads 1-2 mm (0.04-0.08 in)
Molecular sieve A4 (0.4 nm, 4 Å)

CAS No.: 70955-01-0 MDL No.: MFCD00131613

Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR