1-Phenylpyrrolidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

L05239-14 L05239-06
AAAL05239-14EA 535 USD
AAAL05239-14 AAAL05239-06
1-Phenylpyrrolidine 98+%
1-Phenylpyrrolidine

Purity: 98+% CAS No.: 4096-21-3 Molecular Formula: C10H13N Formula Weight: 147.22 Refractive Index: 1.5820 Boiling Point: 133-134°/19mm MDL No.: MFCD00015897

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 55 - 60 °C (0.1 -rr)
Melting Pt: 11 °C
Density: 1.018 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015897
CAS Number: 4096-21-3
EINECS: 223-849-0
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR