1-Chloro-6-iodohexane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05198-09
L05198-18
AAAL05198-09EA
131
USD
AAAL05198-09
AAAL05198-18
1-Chloro-6-iodohexane 97%
1-Chloro-6-iodohexane
Purity: 97%
CAS No.: 34683-73-3
Molecular Formula: Cl(CH2)6I
Formula Weight: 246.52
Storage Temperature: -30°C to -10°C
Refractive Index: 1.5250
Beilstein Reference: 1732408
MDL No.: MFCD00039417
Formula:
C₆H₁₂ClI MW: 246.52 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00039417 CAS Number: 34683-73-3 EINECS: 252-144-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...