(±)-1,2,4-Butanetriol 96%
Supplier: Thermo Scientific Chemicals
L05138-14
L05138-06
AAAL05138-14EA
80.09
USD
AAAL05138-14
AAAL05138-06
(±)-1,2,4-Butanetriol 96%
(±)-1,2,4-Butanetriol
Purity: 96%
CAS No.: 3068-00-6
Molecular Formula: C4H10O3
Formula Weight: 106.12
Flash Point: 188°(370°F)
Refractive Index: 1.4750
Melting Point: -20°
Boiling Point: 190-191°/18mm
Beilstein Reference: 1733456
MDL No.: MFCD00002929
Formula:
C₄H₁₀O₃ Boiling Pt: 190…191 °C/18 mm Melting Pt: –20 °C Density: 1.21 g/cm³ (0 °C) Flash Pt: 188 °C(370 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002929 CAS Number: 3068-00-6 EINECS: 221-323-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...