7-Bromoheptanenitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Bromoheptanonitrile

L05068-06 L05068-14
AAAL05068-06EA 57.1 USD
AAAL05068-06 AAAL05068-14
7-Bromoheptanenitrile 98%
7-Bromoheptanenitrile

Purity: 98% CAS No.: 20965-27-9 Molecular Formula: Br(CH2)6CN Formula Weight: 190.09 Flash Point: 110°(230°F) Refractive Index: 1.4750 Boiling Point: 114-115°/4mm Beilstein Reference: 1748070 UN No.: 3276 MDL No.: MFCD00001982

Formula: BrH₂C(CH₂)₄CH₂CN
MW: 190.08 g/mol
Boiling Pt: 114…115 °C/4 mm
Density: 1.26 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001982
CAS Number: 20965-27-9
EINECS: 244-128-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR