Succinonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butanedinitrile, Succinodinitrile

L05037-36 L05037-22
AAAL05037-36EA 163 USD
AAAL05037-36 AAAL05037-22
Succinonitrile 98%
Succinonitrile

Purity: 98% CAS No.: 110-61-2 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 132°(269°F) Odor: Odorless Boiling Point: 265-267° Beilstein Reference: 1098380 Merck Reference: 14,8872 UN No.: 3439 MDL No.: MFCD00001949

Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 52…56 °C
Density: 1.023 g/cm³ (20 °C)
Flash Pt: 132 °C(269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001949
CAS Number: 110-61-2
EINECS: 203-783-9
ADR: 6.1,III
Merck Index: 12,09041

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR