3-Acetoxyindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate

L04932-03 L04932-06
AAAL04932-03EA 55.8 USD
AAAL04932-03 AAAL04932-06
3-Acetoxyindole 97%
3-Acetoxyindole

Useful for detection of esterases


Purity: 97% CAS No.: 608-08-2 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 128-130° Beilstein Reference: 143086

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00014561
CAS Number: 608-08-2
EINECS: 210-154-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR