4-Bromoheptane 97%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

L04807-06 L04807-14
AAAL04807-06EA 59.5 USD
AAAL04807-06 AAAL04807-14
4-Bromoheptane 97%, Technical Grade
4-Bromoheptane

Purity: 97% CAS No.: 998-93-6 Molecular Formula: C7H15Br Formula Weight: 179.11 Flash Point: 40°(104°F) Refractive Index: 1.4500 Boiling Point: 60-62°/18mm Beilstein Reference: 1697144 UN No.: 1993 MDL No.: MFCD00039182

Formula: H₃C(CH₂)₂BrCH(CH₂)₂CH₃
MW: 179.1 g/mol
Boiling Pt: 60…62 °C/18 mm
Density: 1.1565 g/cm³ (0 °C)
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039182
CAS Number: 998-93-6
EINECS: 213-653-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR