3-Chlorothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Chlorobenzene-1-thiol, 1-Chloro-3-mercaptobenzene, 3-Chlorobenzenethiol, m-Chlorothiophenol

L04806-06 L04806-03 L04806-14
AAAL04806-06EA 51 USD
AAAL04806-06 AAAL04806-03 AAAL04806-14
3-Chlorothiophenol 97%
3-Chlorothiophenol

Purity: 97% CAS No.: 2037-31-2 Molecular Formula: C6H5ClS Formula Weight: 144.62 Flash Point: 90°(194°F) Refractive Index: 1.6021 Boiling Point: 104-105°/35mm Beilstein Reference: 1854087 UN No.: 1760 MDL No.: MFCD00004839

Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 104…105 °C/35 mm
Density: 1.2637 g/cm³ (12.5 °C)
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004839
CAS Number: 2037-31-2
EINECS: 218-010-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR