1-Bromo-4-cyclohexylbenzene 98%

Supplier: Thermo Scientific Chemicals

L04636-14 L04636-06
AAAL04636-14EA 336 USD
AAAL04636-14 AAAL04636-06
1-Bromo-4-cyclohexylbenzene 98%
1-Bromo-4-cyclohexylbenzene

Purity: 98% CAS No.: 25109-28-8 Molecular Formula: C12H15Br Formula Weight: 239.16 Refractive Index: 1.5595 Boiling Point: 90-94°/0.01mm MDL No.: MFCD00019339

Formula: C₁₂H₁₅Br
MW: 239.16 g/mol
Boiling Pt: 90…94 °C (0.01 mmHg)
Density: 1.287
Storage Temperature: Ambient
MDL Number: MFCD00019339
CAS Number: 25109-28-8
EINECS: 246-623-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR