4-Methylpyrocatechol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methylbenzene-1,2-diol, 4-Methylcatechol
L04495-14
L04495-22
AAAL04495-36EA
277.76
USD
AAAL04495-36
AAAL04495-14
AAAL04495-22
4-Methylpyrocatechol 96%
4-Methylpyrocatechol
Purity: 96%
CAS No.: 452-86-8
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 140°(284°F)
Melting Point: 67-69°
Boiling Point: 251°
Beilstein Reference: 636512
MDL No.: MFCD00002205
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 251 °C Melting Pt: 67…69 °C Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002205 CAS Number: 452-86-8 EINECS: 207-214-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...