1-Chlorooctane 99%

Supplier: Thermo Scientific Chemicals
Warning

L04358-AE L04358-AP
AAAL04358-AEEA 24 USD
AAAL04358-AE AAAL04358-AP
1-Chlorooctane 99%
1-Chlorooctane
Formula: C₈H₁₇Cl
Boiling Pt: 182…183 °C
Melting Pt: –58 °C
Density: 0.873
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001022
CAS Number: 111-85-3
EINECS: 203-915-5
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR